missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2-Chloro-6-methylisonicotinate, TCI America™

$33.61 - $99.31

Chemical Identifiers

CAS 3998-90-1
Molecular Formula C8H8ClNO2
Molecular Weight (g/mol) 185.607
MDL Number MFCD04038391
InChI Key BDWMGYZSQKGUFA-UHFFFAOYSA-N
Synonym methyl 2-chloro-6-methylisonicotinate, methyl 2-chloro-6-picoline-4-carboxylate, 2-chloro-6-picoline-4-carboxylic acid methyl ester, 2-chloro-6-methylpyridine-4-carboxylic acid methyl ester, 2-chloro-4-methoxycarbonyl-6-methylpyridine, methyl 6-chloro-2-methylpyridine-4-carboxylate, 2-chloro-6-methylisonicotinic acid methyl ester, 2-chloro-6-methyl pyridine-4-carboxylic acid methyl ester, 4-pyridinecarboxylic acid, 2-chloro-6-methyl-, methyl ester, pubchem17060
PubChem CID 2759833
IUPAC Name methyl 2-chloro-6-methylpyridine-4-carboxylate
SMILES CC1=NC(=CC(=C1)C(=O)OC)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M22395G View Documents TCI America
M22395G
5g Each for $99.31
 
M22391G View Documents TCI America
M22391G
1g Each for $33.61
 
Specifications

Chemical Identifiers

3998-90-1
185.607
BDWMGYZSQKGUFA-UHFFFAOYSA-N
2759833
CC1=NC(=CC(=C1)C(=O)OC)Cl
C8H8ClNO2
MFCD04038391
methyl 2-chloro-6-methylisonicotinate, methyl 2-chloro-6-picoline-4-carboxylate, 2-chloro-6-picoline-4-carboxylic acid methyl ester, 2-chloro-6-methylpyridine-4-carboxylic acid methyl ester, 2-chloro-4-methoxycarbonyl-6-methylpyridine, methyl 6-chloro-2-methylpyridine-4-carboxylate, 2-chloro-6-methylisonicotinic acid methyl ester, 2-chloro-6-methyl pyridine-4-carboxylic acid methyl ester, 4-pyridinecarboxylic acid, 2-chloro-6-methyl-, methyl ester, pubchem17060
methyl 2-chloro-6-methylpyridine-4-carboxylate

Specifications

125°C
60°C
3998-90-1
MFCD04038391
BDWMGYZSQKGUFA-UHFFFAOYSA-N
methyl 2-chloro-6-methylpyridine-4-carboxylate
2759833
Crystal-Powder at 20°C
Methyl 2-Chloro-6-methylisonicotinate
White-Yellow
5g
C8H8ClNO2
methyl 2-chloro-6-methylisonicotinate, methyl 2-chloro-6-picoline-4-carboxylate, 2-chloro-6-picoline-4-carboxylic acid methyl ester, 2-chloro-6-methylpyridine-4-carboxylic acid methyl ester, 2-chloro-4-methoxycarbonyl-6-methylpyridine, methyl 6-chloro-2-methylpyridine-4-carboxylate, 2-chloro-6-methylisonicotinic acid methyl ester, 2-chloro-6-methyl pyridine-4-carboxylic acid methyl ester, 4-pyridinecarboxylic acid, 2-chloro-6-methyl-, methyl ester, pubchem17060
CC1=NC(=CC(=C1)C(=O)OC)Cl
185.607
185.61
≥97.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No