Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-chloro-5-nitrobenzoate, 97%

$34.32 - $109.43

Chemical Identifiers

Chemical Name or Material Methyl 2-chloro-5-nitrobenzoate
CAS 6307-82-0
Molecular Formula C8H6ClNO4
Formula Weight 215.59
MDL Number MFCD00184587
Synonym 2-Chloro-5-nitrobenzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1991309

 
alfa aesar™
A1991309
10g Each for $109.43

AAA1991388

 
alfa aesar™
A1991388
2.5g Each for $34.32
Description & Specifications

Specifications

Formula Weight 215.59
Chemical Name or Material Methyl 2-chloro-5-nitrobenzoate
Assay 97%
Molecular Formula C8H6ClNO4
MDL Number MFCD00184587
CAS 6307-82-0
Synonym 2-Chloro-5-nitrobenzoic acid methyl ester
Health Hazard 1 Warning
Melting Point 70° to 72°C
Quantity 10g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
Methyl 2-chloro-5-nitrobenzoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.