missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate, 97%, Acros Organics™

Manufacturer:  Acros Organics AC448690250

 View more versions of this product

Catalog No. AC448690250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Methyl 2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate
Authentic
25g
C7H4ClF3N2O2
VLUBJYUOPNGBOQ-UHFFFAOYSA-N
methyl 2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate
3696469
97%
110°C
56.0°C (0.2 mmHg)
175137-27-6
methyl 2-chloro-4-trifluoromethyl pyrimidine-5-carboxylate, methyl 2-chloro-4-trifluoromethyl-5-pyrimidinecarboxylate, methylchlorotrifluoromethylpyrimidinecarboxylate, methyl-2-chloro-4-trifluoromethyl pyrimidine-5-carboxylate, acmc-20aifi, pubchem10174, 2-chloro-4-trifluoromethyl-5-carbomethoxypyrimidine, methyl 2-chloro-4-trifluoromethyl pyrimidine-5-ca, 2-chloro-5-methoxycarbonyl-4-trifluoromethylpyrimidine, 2-chloro-5-methoxycarbonyl-4-trifluoromethyl pyrimidine
COC(=O)C1=CN=C(N=C1C(F)(F)F)Cl
240.566
240.57

Chemical Identifiers

175137-27-6
240.566
methyl 2-chloro-4-trifluoromethyl pyrimidine-5-carboxylate, methyl 2-chloro-4-trifluoromethyl-5-pyrimidinecarboxylate, methylchlorotrifluoromethylpyrimidinecarboxylate, methyl-2-chloro-4-trifluoromethyl pyrimidine-5-carboxylate, acmc-20aifi, pubchem10174, 2-chloro-4-trifluoromethyl-5-carbomethoxypyrimidine, methyl 2-chloro-4-trifluoromethyl pyrimidine-5-ca, 2-chloro-5-methoxycarbonyl-4-trifluoromethylpyrimidine, 2-chloro-5-methoxycarbonyl-4-trifluoromethyl pyrimidine
methyl 2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate
C7H4ClF3N2O2
VLUBJYUOPNGBOQ-UHFFFAOYSA-N
3696469
COC(=O)C1=CN=C(N=C1C(F)(F)F)Cl
Safety and Handling

Safety and Handling

GHS H Statement Harmful if inhaled. Causes serious eye irritation. Causes skin irritation. May cause respiratory irritation. Harmful in contact with skin. Harmful if swallowed.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

Warning

SDS Documents