Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-chloro-4-iodonicotinate, 97%

$155.47 - $155.47

Chemical Identifiers

Chemical Name or Material Methyl 2-chloro-4-iodonicotinate
CAS 185041-05-8
Molecular Formula C7H5ClINO2
Formula Weight 297.48
MDL Number MFCD04039871
Synonym 2-Chloro-4-iodonicotinic acid methyl ester; Methyl 2-chloro-4-iodopyridine-3-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5731903

 
alfa aesar™
H5731903
1g Each for $155.47

AAH57319MD

 
alfa aesar™
H57319MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 297.48
Chemical Name or Material Methyl 2-chloro-4-iodonicotinate
Assay 97%
Molecular Formula C7H5ClINO2
MDL Number MFCD04039871
CAS 185041-05-8
Synonym 2-Chloro-4-iodonicotinic acid methyl ester; Methyl 2-chloro-4-iodopyridine-3-carboxylate
Melting Point 81° to 85°C
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.