missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2-Chloro-4-fluorobenzoate, TCI America™

$70.28 - $213.89

Chemical Identifiers

CAS 85953-29-3
Molecular Formula C8H6ClFO2
Molecular Weight (g/mol) 188.582
MDL Number MFCD00173937
InChI Key YZDLPZNWBRBZMZ-UHFFFAOYSA-N
Synonym 2-chloro-4-fluorobenzoic acid methyl ester, methyl 2-chloro-4-fluorobenzenecarboxylate, benzoic acid, 2-chloro-4-fluoro-, methyl ester, 2-chloro-4-fluoro-benzoic acid methyl ester, pubchem18760, rarechem al bf 0289, acmc-209q7x, ksc495c1p, methyl 2-chloro-4-fluorob..., methy 2-chloro-4-fluorobenzoate
PubChem CID 2763551
IUPAC Name methyl 2-chloro-4-fluorobenzoate
SMILES COC(=O)C1=C(C=C(C=C1)F)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M31025G View Documents TCI America
M31025G
5g Each for $70.28
 
M310225G View Documents TCI America
M310225G
25g Each for $213.89
 
Specifications

Chemical Identifiers

85953-29-3
188.582
YZDLPZNWBRBZMZ-UHFFFAOYSA-N
2763551
COC(=O)C1=C(C=C(C=C1)F)Cl
C8H6ClFO2
MFCD00173937
2-chloro-4-fluorobenzoic acid methyl ester, methyl 2-chloro-4-fluorobenzenecarboxylate, benzoic acid, 2-chloro-4-fluoro-, methyl ester, 2-chloro-4-fluoro-benzoic acid methyl ester, pubchem18760, rarechem al bf 0289, acmc-209q7x, ksc495c1p, methyl 2-chloro-4-fluorob..., methy 2-chloro-4-fluorobenzoate
methyl 2-chloro-4-fluorobenzoate

Specifications

105°C
5g
C8H6ClFO2
2-chloro-4-fluorobenzoic acid methyl ester, methyl 2-chloro-4-fluorobenzenecarboxylate, benzoic acid, 2-chloro-4-fluoro-, methyl ester, 2-chloro-4-fluoro-benzoic acid methyl ester, pubchem18760, rarechem al bf 0289, acmc-209q7x, ksc495c1p, methyl 2-chloro-4-fluorob..., methy 2-chloro-4-fluorobenzoate
COC(=O)C1=C(C=C(C=C1)F)Cl
188.582
188.58
≥98.0% (GC)
Yellow
85953-29-3
MFCD00173937
YZDLPZNWBRBZMZ-UHFFFAOYSA-N
methyl 2-chloro-4-fluorobenzoate
2763551
Clear Liquid at 20°C
Methyl 2-Chloro-4-fluorobenzoate
Safety and Handling

Safety and Handling

TSCA : No