missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-chloro-4-fluorobenzoate, 97%

$93.54 - $355.19

Chemical Identifiers

CAS 85953-29-3
Molecular Formula C8H6ClFO2
Molecular Weight (g/mol) 188.582
MDL Number MFCD00173937
InChI Key YZDLPZNWBRBZMZ-UHFFFAOYSA-N
Synonym 2-chloro-4-fluoro-benzoic acid methyl ester
PubChem CID 2763551
IUPAC Name methyl 2-chloro-4-fluorobenzoate
SMILES COC(=O)C1=C(C=C(C=C1)F)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1834203 View Documents Alfa Aesar™
A1834203
1g Each for $93.54
 
AAA1834206 View Documents Alfa Aesar™
A1834206
5g Each for $355.19
 
Specifications

Chemical Identifiers

85953-29-3
188.582
YZDLPZNWBRBZMZ-UHFFFAOYSA-N
2763551
COC(=O)C1=C(C=C(C=C1)F)Cl
C8H6ClFO2
MFCD00173937
2-chloro-4-fluoro-benzoic acid methyl ester
methyl 2-chloro-4-fluorobenzoate

Specifications

103°C to 105°C (15mmHg)
85953-29-3
MFCD00173937
YZDLPZNWBRBZMZ-UHFFFAOYSA-N
methyl 2-chloro-4-fluorobenzoate
2763551
97%
1g
C8H6ClFO2
2-chloro-4-fluoro-benzoic acid methyl ester
COC(=O)C1=C(C=C(C=C1)F)Cl
188.582
188.59
Methyl 2-chloro-4-fluorobenzoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning