Please login to your online account to display your discounted pricing

Methyl 2-chloro-2,2-difluoroacetate, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyl 2-chloro-2, 2-difluoroacetate
Name Note 99%
CAS 1514-87-0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C3H3ClF2O2
Linear Formula ClCFCOCH3
Formula Weight 144.51
MDL Number MFCD00000775
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC308010050

  SDS
thermo fisher scientific
308010050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC308010250

  SDS
thermo fisher scientific
308010250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Liquid
Formula Weight 144.51
Quantity 5g
Color Clear colorless
EINECS Number 216-154-9
TSCA TSCA
Density 1.37g/mL
Chemical Name or Material Methyl 2-chloro-2, 2-difluoroacetate
Refractive Index 1.3480 to 1.3500 (20°C, 589nm)
Name Note 99%
Assay Percent Range 99%
Molecular Formula C3H3ClF2O2
Linear Formula ClCFCOCH3
MDL Number MFCD00000775
CAS 1514-87-0
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapor.