missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2-Bromothiazole-5-carboxylate, TCI America™

$117.64 - $348.34

Chemical Identifiers

CAS 54045-74-8
Molecular Formula C5H4BrNO2S
Molecular Weight (g/mol) 222.056
MDL Number MFCD03788565
InChI Key HNLVIKMXFBRZDF-UHFFFAOYSA-N
Synonym methyl 2-bromothiazole-5-carboxylate, methyl 2-bromo-5-thiazole carboxylate, methyl2-bromothiazole-5-carboxylate, 5-thiazolecarboxylic acid, 2-bromo-, methyl ester, 2-bromo-thiazole-5-carboxylic acid methyl ester, 2-bromothiazole-5-carboxylic acid methyl ester, 2-bromo-1,3-thiazole-5-carboxylic acid methyl ester, pubchem17197, acmc-1apjm, ksc494s7p
PubChem CID 2773502
IUPAC Name methyl 2-bromo-1,3-thiazole-5-carboxylate
SMILES COC(=O)C1=CN=C(S1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M26695G View Documents TCI America
M26695G
5g Each for $348.34
 
M26691G View Documents TCI America
M26691G
1g Each for $117.64
 
Specifications

Chemical Identifiers

54045-74-8
222.056
HNLVIKMXFBRZDF-UHFFFAOYSA-N
2773502
COC(=O)C1=CN=C(S1)Br
C5H4BrNO2S
MFCD03788565
methyl 2-bromothiazole-5-carboxylate, methyl 2-bromo-5-thiazole carboxylate, methyl2-bromothiazole-5-carboxylate, 5-thiazolecarboxylic acid, 2-bromo-, methyl ester, 2-bromo-thiazole-5-carboxylic acid methyl ester, 2-bromothiazole-5-carboxylic acid methyl ester, 2-bromo-1,3-thiazole-5-carboxylic acid methyl ester, pubchem17197, acmc-1apjm, ksc494s7p
methyl 2-bromo-1,3-thiazole-5-carboxylate

Specifications

Undesignated
5g
C5H4BrNO2S
methyl 2-bromothiazole-5-carboxylate, methyl 2-bromo-5-thiazole carboxylate, methyl2-bromothiazole-5-carboxylate, 5-thiazolecarboxylic acid, 2-bromo-, methyl ester, 2-bromo-thiazole-5-carboxylic acid methyl ester, 2-bromothiazole-5-carboxylic acid methyl ester, 2-bromo-1,3-thiazole-5-carboxylic acid methyl ester, pubchem17197, acmc-1apjm, ksc494s7p
COC(=O)C1=CN=C(S1)Br
222.056
222.06
≥95.0% (GC)
62°C
54045-74-8
MFCD03788565
HNLVIKMXFBRZDF-UHFFFAOYSA-N
methyl 2-bromo-1,3-thiazole-5-carboxylate
2773502
Crystal-Powder at 20°C
Methyl 2-Bromothiazole-5-carboxylate
Safety and Handling

Safety and Handling

TSCA : No