Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-bromothiazole-5-carboxylate, 98%

$262.18 - $798.58

Chemical Identifiers

Chemical Name or Material Methyl 2-bromothiazole-5-carboxylate
CAS 54045-74-8
Molecular Formula C5H4BrNO2S
Formula Weight 222.05
MDL Number MFCD03788565
Synonym 2-Bromothiazole-5-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2741003

  SDS
alfa aesar™
H2741003
1g Each for $262.18

AAH2741006

  SDS
alfa aesar™
H2741006
5g Each for $798.58
Description & Specifications

Specifications

Formula Weight 222.05
Chemical Name or Material Methyl 2-bromothiazole-5-carboxylate
Assay 98%
Molecular Formula C5H4BrNO2S
MDL Number MFCD03788565
CAS 54045-74-8
Synonym 2-Bromothiazole-5-carboxylic acid methyl ester
Health Hazard 1 Warning
Melting Point 61° to 64°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.