missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2-Bromonicotinate, TCI America™

$76.39 - $255.14

Chemical Identifiers

CAS 52718-95-3
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD07367915
InChI Key RAFFKXWNTXTTFO-UHFFFAOYSA-N
Synonym 2-Bromonicotinic Acid Methyl Ester, Methyl 2-Bromopyridine-3-carboxylate, 2-Bromopyridine-3-carboxylic Acid Methyl Ester
PubChem CID 12349191
IUPAC Name methyl 2-bromopyridine-3-carboxylate
SMILES COC(=O)C1=C(N=CC=C1)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M24491G View Documents TCI America
M24491G
1g Each for $76.39
 
M24495G View Documents TCI America
M24495G
5g Each for $255.14
 
Specifications

Chemical Identifiers

52718-95-3
216.034
RAFFKXWNTXTTFO-UHFFFAOYSA-N
12349191
COC(=O)C1=C(N=CC=C1)Br
C7H6BrNO2
MFCD07367915
2-Bromonicotinic Acid Methyl Ester, Methyl 2-Bromopyridine-3-carboxylate, 2-Bromopyridine-3-carboxylic Acid Methyl Ester
methyl 2-bromopyridine-3-carboxylate

Specifications

Undesignated
52718-95-3
MFCD07367915
RAFFKXWNTXTTFO-UHFFFAOYSA-N
methyl 2-bromopyridine-3-carboxylate
12349191
Crystal-Lump at 20°C
Methyl 2-Bromonicotinate
1g
C7H6BrNO2
2-Bromonicotinic Acid Methyl Ester, Methyl 2-Bromopyridine-3-carboxylate, 2-Bromopyridine-3-carboxylic Acid Methyl Ester
COC(=O)C1=C(N=CC=C1)Br
216.034
216.03
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No