Please login to your online account to display your discounted pricing

Methyl 2-(bromomethyl)acrylate, ACROS Organics™


Quantity 250mg

Manufacturer: thermo fisher scientific  AC367632500

 View more versions of this product

Catalog No. AC367632500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Methyl 2-(bromomethyl)acrylate
CAS 4224-69-5
Density 1.489g/mL
Infrared Spectrum Authentic
Physical Form Liquid
Refractive Index 1.4870 to 1.4890 (20°C, 589nm)
Assay ≥95% (GC)
Assay Percent Range 96%
Molecular Formula C5H7BrO2
Formula Weight 179.01
Color Clear colorless to slightly brownish
Quantity 250mg

Specifications

Chemical Name or Material Methyl 2-(bromomethyl)acrylate
Assay ≥95% (GC)
Assay Percent Range 96%
Molecular Formula C5H7BrO2
Formula Weight 179.01