Learn More
Methyl 2-(bromomethyl)acrylate, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 367630250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Methyl 2-(bromomethyl)acrylate | |
1.4890g/mL | |
78°C | |
95% min. (GC) | |
C5H7BrO2 | |
MFCD00011697 | |
1.489 | |
CFTUQSLVERGMHL-UHFFFAOYSA-N | |
methyl 2-(bromomethyl)prop-2-enoate | |
521093 | |
96% |
4224-69-5 | |
35.0°C to 37.0°C (13.0mmHg) | |
Authentic | |
Glass bottle | |
1.4870 to 1.4890 | |
25g | |
methyl 2-bromomethyl acrylate, methyl 2-bromomethyl prop-2-enoate, methyl 2-bromomethyl-2-propenoate, methyl alpha-bromomethyl acrylate, 2-bromomethyl methyl acrylate, 2-propenoic acid, 2-bromomethyl-, methyl ester, 2-bromomethyl acrylic acid methyl ester, acrylic acid, 2-bromomethyl-, methyl ester, zlchem 195, pubchem17371 | |
COC(=O)C(=C)CBr | |
179.01 | |
179.01 |
Chemical Identifiers
4224-69-5 | |
179.01 | |
CFTUQSLVERGMHL-UHFFFAOYSA-N | |
521093 | |
COC(=O)C(=C)CBr |
C5H7BrO2 | |
MFCD00011697 | |
methyl 2-bromomethyl acrylate, methyl 2-bromomethyl prop-2-enoate, methyl 2-bromomethyl-2-propenoate, methyl alpha-bromomethyl acrylate, 2-bromomethyl methyl acrylate, 2-propenoic acid, 2-bromomethyl-, methyl ester, 2-bromomethyl acrylic acid methyl ester, acrylic acid, 2-bromomethyl-, methyl ester, zlchem 195, pubchem17371 | |
methyl 2-(bromomethyl)prop-2-enoate |
Safety and Handling
GHS H Statement
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
GHS Signal Word: Danger
RUO â Research Use Only