missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2-(Bromomethyl)-3-nitrobenzoate 98.0+%, TCI America™

$148.13 - $487.17

Chemical Identifiers

CAS 98475-07-1
Molecular Formula C9H8BrNO4
Molecular Weight (g/mol) 274.07
MDL Number MFCD04114315
InChI Key FCGIVHSBEKGQMZ-UHFFFAOYSA-N
Synonym 2-bromomethyl-3-nitro benzoic acid methyl ester, 2-bromomethyl-3-nitrobenzoic acid methyl ester, benzoic acid, 2-bromomethyl-3-nitro-, methyl ester, methyl 2-bromomethyl-3-nitro-benzoate, methyl 2-bromomethyl-3-nitrobenzenecarboxylate, methyl 2-bromomethyl-3-nitrobenzoate, methyl2-bromomethyl-3-nitrobenzoate, pubchem19169, zlchem 474
PubChem CID 1487795
IUPAC Name methyl 2-(bromomethyl)-3-nitrobenzoate
SMILES COC(=O)C1=C(C(=CC=C1)[N+](=O)[O-])CBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M24221G View Documents Tci America
M24221G
1g Each for $148.13
 
M24225G View Documents Tci America
M24225G
5g Each for $487.17
 
Specifications

Chemical Identifiers

98475-07-1
274.07
FCGIVHSBEKGQMZ-UHFFFAOYSA-N
1487795
COC(=O)C1=C(C(=CC=C1)[N+](=O)[O-])CBr
C9H8BrNO4
MFCD04114315
2-bromomethyl-3-nitro benzoic acid methyl ester, 2-bromomethyl-3-nitrobenzoic acid methyl ester, benzoic acid, 2-bromomethyl-3-nitro-, methyl ester, methyl 2-bromomethyl-3-nitro-benzoate, methyl 2-bromomethyl-3-nitrobenzenecarboxylate, methyl 2-bromomethyl-3-nitrobenzoate, methyl2-bromomethyl-3-nitrobenzoate, pubchem19169, zlchem 474
methyl 2-(bromomethyl)-3-nitrobenzoate

Specifications

Yellow
1g
C9H8BrNO4
3261
FCGIVHSBEKGQMZ-UHFFFAOYSA-N
methyl 2-(bromomethyl)-3-nitrobenzoate
1487795
Crystal-Powder at 20°C
Methyl 2-(Bromomethyl)-3-nitrobenzoate
73°C
98475-07-1
MFCD04114315
2-bromomethyl-3-nitro benzoic acid methyl ester, 2-bromomethyl-3-nitrobenzoic acid methyl ester, benzoic acid, 2-bromomethyl-3-nitro-, methyl ester, methyl 2-bromomethyl-3-nitro-benzoate, methyl 2-bromomethyl-3-nitrobenzenecarboxylate, methyl 2-bromomethyl-3-nitrobenzoate, methyl2-bromomethyl-3-nitrobenzoate, pubchem19169, zlchem 474
COC(=O)C1=C(C(=CC=C1)[N+](=O)[O-])CBr
274.07
274.07
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No