Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-bromomethyl-3-nitrobenzoate, 95%

Chemical Identifiers

Chemical Name or Material Methyl 2-bromomethyl-3-nitrobenzoate
CAS 98475-07-1
Formula Weight 274.07
MDL Number MFCD04114315
Beilstein 7811923
Synonym 2-Bromomethyl-3-nitrobenzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6535103

 
alfa aesar™
H6535103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH65351MD

 
alfa aesar™
H65351MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 274.07
Chemical Name or Material Methyl 2-bromomethyl-3-nitrobenzoate
MDL Number MFCD04114315
CAS 98475-07-1
Assay 95%
Physical Form Solid
Synonym 2-Bromomethyl-3-nitrobenzoic acid methyl ester
Quantity 1g
Beilstein 7811923