missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-bromohexanoate, 98%

$40.41 - $117.82

Chemical Identifiers

CAS 5445-19-2
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.083
MDL Number MFCD00045038
InChI Key YGLPDRIMFIXNBI-UHFFFAOYSA-N
Synonym 2-bromohexanoic acid methyl ester, hexanoic acid, 2-bromo-, methyl ester, methyl 2-bromocaproate, methyl 2-bromanylhexanoate, methyl bromohexanoate, hexanoic acid, 2-bromo-, methyl ester, s, methyl-2-bromohexanoate, acmc-20mk9n, ksc490a4j, methyl-2-bromocaproate
PubChem CID 95578
IUPAC Name methyl 2-bromohexanoate
SMILES CCCCC(C(=O)OC)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60165AC View Documents Alfa Aesar™
H60165AC
25mL Each for $40.41
 
AAH60165AE View Documents Alfa Aesar™
H60165AE
100mL Each for $117.82
 
Specifications

Chemical Identifiers

5445-19-2
209.083
YGLPDRIMFIXNBI-UHFFFAOYSA-N
95578
CCCCC(C(=O)OC)Br
C7H13BrO2
MFCD00045038
2-bromohexanoic acid methyl ester, hexanoic acid, 2-bromo-, methyl ester, methyl 2-bromocaproate, methyl 2-bromanylhexanoate, methyl bromohexanoate, hexanoic acid, 2-bromo-, methyl ester, s, methyl-2-bromohexanoate, acmc-20mk9n, ksc490a4j, methyl-2-bromocaproate
methyl 2-bromohexanoate

Specifications

69°C (156°F)
5445-19-2
CH3(CH2)3CHBrCOOCH3
1722052
YGLPDRIMFIXNBI-UHFFFAOYSA-N
methyl 2-bromohexanoate
95578
98%
25mL
C7H13BrO2
MFCD00045038
2-bromohexanoic acid methyl ester, hexanoic acid, 2-bromo-, methyl ester, methyl 2-bromocaproate, methyl 2-bromanylhexanoate, methyl bromohexanoate, hexanoic acid, 2-bromo-, methyl ester, s, methyl-2-bromohexanoate, acmc-20mk9n, ksc490a4j, methyl-2-bromocaproate
CCCCC(C(=O)OC)Br
209.083
209.08
Methyl 2-bromohexanoate
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA