Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-bromobutyrate, 97%

Chemical Identifiers

Chemical Name or Material Methyl 2-bromobutyrate
CAS 3196-15-4
Molecular Formula C5H9BrO2
Formula Weight 181.04
MDL Number MFCD00009666
UN Number UN1760
Beilstein 1720826
Synonym 2-Bromobutyric acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA194680B

  SDS
alfa aesar™
A194680B
1000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 181.04
CAS 3196-15-4
Molecular Formula C5H9BrO2
Flash Point 68°C (154°F)
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Assay 97%
Boiling Point 137° to 138°C (50mmHg)
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
MDL Number MFCD00009666
Health Hazard 1 Danger
Quantity 1000g
Chemical Name or Material Methyl 2-bromobutyrate
Synonym 2-Bromobutyric acid methyl ester
UN Number UN1760
Beilstein 1720826
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA

Applications
Methyl 2-bromobutyrate is used in the synthesis of racemic indene ester via Reformatsky reaction with 5-methoxy-1-indanone. (E)-2-methoxymethylene-butyric acid methyl ester via reformatsky reaction with α,α-dichloromethyl methyl ether in the presence of zinc dust.

Solubility
Soluble in alcohol and ether, slightly soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.