missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-bromobenzoate, 99%

$43.73 - $695.76

Chemical Identifiers

CAS 610-94-6
Molecular Formula C8H7BrO2
Formula Weight 215.05
MDL Number MFCD00016328
Beilstein 1862501
Synonym 2-Bromobenzoic acid methyl ester
Chemical Name or Material Methyl 2-bromobenzoate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1243514 SDS Alfa Aesar™
A1243514
25g Each for $43.73
AAA1243522 SDS Alfa Aesar™
A1243522
100g Each for $160.08
AAA1243536 SDS Alfa Aesar™
A1243536
500g Each for $695.76
Specifications

Description

Applications
Methyl 2-bromobenzoate under Heck reaction with acrylonitrile results in lactonization to give the benzofuran-2-one 3-(cyanomethyl)-2-coumaranone. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals

Solubility
Insoluble in water.

Notes
Store in a cool place. Keep away from oxidizing agents.
Description & Specifications

Chemical Identifiers

610-94-6
215.05
1862501
Methyl 2-bromobenzoate
C8H7BrO2
MFCD00016328
2-Bromobenzoic acid methyl ester

Specifications

610-94-6
215.05
1862501
252°C
25g
99%
C8H7BrO2
MFCD00016328
2-Bromobenzoic acid methyl ester
>110°C (230°F)
Methyl 2-bromobenzoate

Safety and Handling

TSCA : TSCA