Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-bromo-6-fluorophenylacetate, 96%

Chemical Identifiers

Chemical Name or Material Methyl 2-bromo-6-fluorophenylacetate
Formula Weight 247.06
Synonym 2-Bromo-6-fluorophenylacetic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6246503

 
alfa aesar™
H6246503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62465MD

 
alfa aesar™
H62465MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6246506

 
alfa aesar™
H6246506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 247.06
Chemical Name or Material Methyl 2-bromo-6-fluorophenylacetate
Physical Form Solid
Assay 96%
Synonym 2-Bromo-6-fluorophenylacetic acid methyl ester
Quantity 1g