missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2-bromo-5-nitrobenzoate, 98%, ACROS Organics™

Chemical Identifiers

CAS 6942-36-5
Assay Percent Range 98%
Molecular Formula C8 H6 Br N O4
Formula Weight 260.04
MDL Number MFCD00010867
Chemical Name or Material Methyl 2-bromo-5-nitrobenzoate
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC342370250 SDS Thermo Fisher Scientific
34237-0250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

6942-36-5
C8 H6 Br N O4
MFCD00010867
≥97.5%
98%
260.04
Methyl 2-bromo-5-nitrobenzoate

Specifications

6942-36-5
Glass bottle
98%
260.04
79° to 81°C
25g
≥97.5%
Authentic
Crystalline Powder
C8 H6 Br N O4
MFCD00010867
Yellow
Methyl 2-bromo-5-nitrobenzoate