missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 2-Bromo-3-methylbenzoate 98.0+%, TCI America™
Supplier: TCI America M25465G
Specifications
Methyl 2-Bromo-3-methylbenzoate | |
Yellow | |
MFCD00144762 | |
2-bromo-3-methyl-benzoic acid methyl ester, methyl2-bromo-3-methylbenzoate, benzoic acid, 2-bromo-3-methyl-, methyl ester, 2-bromo-3-methylbenzoic acid methyl ester, methyl 2-bromo-3-methyl-benzoate, pubchem3966, acmc-1c3gk, methyl 2-bromo-m-toluate, intermediates-zcf02010, ksc495c9h | |
COC(=O)C1=C(Br)C(C)=CC=C1 | |
229.07 | |
229.07 | |
Liquid |
131001-86-0 | |
C9H9BrO2 | |
5 g | |
QAOFGUXVDAZKBW-UHFFFAOYSA-N | |
methyl 2-bromo-3-methylbenzoate | |
10823279 | |
≥98.0% (GC) |
Chemical Identifiers
131001-86-0 | |
229.07 | |
QAOFGUXVDAZKBW-UHFFFAOYSA-N | |
10823279 | |
COC(=O)C1=C(Br)C(C)=CC=C1 |
C9H9BrO2 | |
MFCD00144762 | |
2-bromo-3-methyl-benzoic acid methyl ester, methyl2-bromo-3-methylbenzoate, benzoic acid, 2-bromo-3-methyl-, methyl ester, 2-bromo-3-methylbenzoic acid methyl ester, methyl 2-bromo-3-methyl-benzoate, pubchem3966, acmc-1c3gk, methyl 2-bromo-m-toluate, intermediates-zcf02010, ksc495c9h | |
methyl 2-bromo-3-methylbenzoate |
Safety and Handling
TSCA : No