missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(±)-Methyl 2-benzyloxycarbonylamino-2-(dimethoxyphosphinyl)acetate, 95%, ACROS Organics™

Chemical Identifiers

CAS 88568-95-0
Molecular Formula C13H18NO7P
Molecular Weight (g/mol) 331.26
MDL Number MFCD00043304
InChI Key GSYSFVSGPABNNL-UHFFFAOYSA-N
Synonym +/--benzyloxycarbonyl-alpha-phosphonoglycine trimethyl ester
PubChem CID 2734718
IUPAC Name methyl 2-dimethoxyphosphoryl-2-(phenylmethoxycarbonylamino)acetate
SMILES COC(=O)C(NC(=O)OCC1=CC=CC=C1)P(=O)(OC)OC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC370040050 SDS View Documents Thermo Fisher Scientific
AC370040050
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

for C-C Bond Formation (Olefination)
Specifications

Chemical Identifiers

88568-95-0
331.26
GSYSFVSGPABNNL-UHFFFAOYSA-N
2734718
COC(=O)C(NC(=O)OCC1=CC=CC=C1)P(=O)(OC)OC
C13H18NO7P
MFCD00043304
+/--benzyloxycarbonyl-alpha-phosphonoglycine trimethyl ester
methyl 2-dimethoxyphosphoryl-2-(phenylmethoxycarbonylamino)acetate

Specifications

Authentic
White
5g
94.0
94% min. (HPLC)
MFCD00043304
GSYSFVSGPABNNL-UHFFFAOYSA-N
methyl 2-dimethoxyphosphoryl-2-(phenylmethoxycarbonylamino)acetate
2734718
Crystalline Powder
(±)-Methyl 2-benzyloxycarbonylamino-2-(dimethoxyphosphinyl)acetate, 95%
Glass bottle
77.0°C to 80.0°C
88568-95-0
100.0
C13H18NO7P
+/--benzyloxycarbonyl-alpha-phosphonoglycine trimethyl ester
COC(=O)C(NC(=O)OCC1=CC=CC=C1)P(=O)(OC)OC
331.26
331.26
95%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning