Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-benzyloxybenzoate, 98%

$51.88 - $492.57

Chemical Identifiers

Chemical Name or Material Methyl 2-benzyloxybenzoate
CAS 55142-16-0
Molecular Formula C15H14O3
Formula Weight 242.27
MDL Number MFCD01099045
Synonym 2-Benzyloxybenzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2330906

 
alfa aesar™
B2330906
5g Each for $51.88

AAB2330922

 
alfa aesar™
B2330922
100g Each for $492.57
Description & Specifications

Specifications

Formula Weight 242.27
Chemical Name or Material Methyl 2-benzyloxybenzoate
Assay 98%
Molecular Formula C15H14O3
MDL Number MFCD01099045
CAS 55142-16-0
Synonym 2-Benzyloxybenzoic acid methyl ester
Melting Point 45° to 48°C
Quantity 5g