missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-aminothiophene-3-carboxylate, 97%

Manufacturer:  Alfa Aesar™ H5548203

 View more versions of this product

Catalog No. AAH5548203



Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Notes
Store at room temperature. Avoid strong oxidizing agents.
Specifications

Specifications

Methyl 2-aminothiophene-3-carboxylate
1g
C6H7NO2S
2-amino-3-methoxycarbonylthiophene
COC(=O)C1=C(SC=C1)N
157.187
157.19
76°C to 81°C
4651-81-4
MFCD00159547
DGGJQLCAYQCPDD-UHFFFAOYSA-N
methyl 2-aminothiophene-3-carboxylate
78381
97%

Chemical Identifiers

4651-81-4
157.187
DGGJQLCAYQCPDD-UHFFFAOYSA-N
78381
COC(=O)C1=C(SC=C1)N
C6H7NO2S
MFCD00159547
2-amino-3-methoxycarbonylthiophene
methyl 2-aminothiophene-3-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents