missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2-Aminothiazole-5-carboxylate, TCI America™

$68.75 - $232.23

Chemical Identifiers

CAS 6633-61-0
Molecular Formula C5H6N2O2S
Molecular Weight (g/mol) 158.175
MDL Number MFCD03788562
InChI Key UJNNCGWBDJHCEM-UHFFFAOYSA-N
Synonym 2-Aminothiazole-5-carboxylic Acid Methyl Ester
PubChem CID 238005
IUPAC Name methyl 2-amino-1,3-thiazole-5-carboxylate
SMILES COC(=O)C1=CN=C(S1)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M25981G View Documents TCI America
M25981G
1g Each for $68.75
 
M25985G View Documents TCI America
M25985G
5g Each for $232.23
 
Specifications

Chemical Identifiers

6633-61-0
158.175
UJNNCGWBDJHCEM-UHFFFAOYSA-N
238005
COC(=O)C1=CN=C(S1)N
C5H6N2O2S
MFCD03788562
2-Aminothiazole-5-carboxylic Acid Methyl Ester
methyl 2-amino-1,3-thiazole-5-carboxylate

Specifications

Yellow
1g
C5H6N2O2S
2-Aminothiazole-5-carboxylic Acid Methyl Ester
COC(=O)C1=CN=C(S1)N
158.175
158.18
≥98.0% (GC)
191°C
6633-61-0
MFCD03788562
UJNNCGWBDJHCEM-UHFFFAOYSA-N
methyl 2-amino-1,3-thiazole-5-carboxylate
238005
Crystal-Powder at 20°C
Methyl 2-Aminothiazole-5-carboxylate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator