missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2-Aminothiazole-4-carboxylate, TCI America™

$113.06 - $337.64

Chemical Identifiers

CAS 118452-04-3
Molecular Formula C5H6N2O2S
Molecular Weight (g/mol) 158.175
MDL Number MFCD00622441
InChI Key WYVZZWKIKAKUKV-UHFFFAOYSA-N
Synonym 2-Aminothiazole-4-carboxylic Acid Methyl Ester
PubChem CID 840099
IUPAC Name methyl 2-amino-1,3-thiazole-4-carboxylate
SMILES COC(=O)C1=CSC(=N1)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M25811G View Documents TCI America
M25811G
1g Each for $113.06
 
M25815G View Documents TCI America
M25815G
5g Each for $337.64
 
Specifications

Chemical Identifiers

118452-04-3
158.175
WYVZZWKIKAKUKV-UHFFFAOYSA-N
840099
COC(=O)C1=CSC(=N1)N
C5H6N2O2S
MFCD00622441
2-Aminothiazole-4-carboxylic Acid Methyl Ester
methyl 2-amino-1,3-thiazole-4-carboxylate

Specifications

Undesignated
1g
C5H6N2O2S
2-Aminothiazole-4-carboxylic Acid Methyl Ester
COC(=O)C1=CSC(=N1)N
158.175
158.18
≥98.0% (GC,T)
174°C
118452-04-3
MFCD00622441
WYVZZWKIKAKUKV-UHFFFAOYSA-N
methyl 2-amino-1,3-thiazole-4-carboxylate
840099
Crystal-Powder at 20°C
Methyl 2-Aminothiazole-4-carboxylate
Safety and Handling

Safety and Handling

TSCA : No