Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-aminonicotinate, 97%

$137.62 - $503.34

Chemical Identifiers

Chemical Name or Material Methyl 2-aminonicotinate
CAS 14667-47-1
Molecular Formula C7H8N2O2
Formula Weight 152.15
MDL Number MFCD00128872
Synonym 2-Aminonicotinic acid methyl ester; Methyl pyridine-3-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5001303

 
alfa aesar™
H5001303
1g Each for $137.62

AAH5001306

 
alfa aesar™
H5001306
5g Each for $503.34
Description & Specifications

Specifications

Formula Weight 152.15
Chemical Name or Material Methyl 2-aminonicotinate
Assay 97%
Molecular Formula C7H8N2O2
MDL Number MFCD00128872
CAS 14667-47-1
Synonym 2-Aminonicotinic acid methyl ester; Methyl pyridine-3-carboxylate
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.