Learn More
Methyl 2-amino-5-methoxybenzoate, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H54704MD
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Slightly Soluble (1.1 g/L) (25°C) in water.
Notes
Store at room temperature. Keep away from oxidizing agents and acids. Moisture sensitive.
Specifications
Methyl 2-amino-5-methoxybenzoate | |
32°C to 33°C | |
MFCD09038482 | |
benzoic acid, 2-amino-5-methoxy-, methyl ester, methyl2-amino-5-methoxybenzoate, 2-amino-5-methoxybenzoic acid methyl ester, methyl 5-methoxyanthranilate, benzoic acid,2-amino-5-methoxy-, methyl ester, methyl 6-amino-m-anisate, acmc-1cnv1, intermediates-zcf02665, 2-amino-5-methoxybenzoic acid,methyl ester, 2-amino-5-methoxy-benzoic acid methyl ester | |
MOVBJUGHBJJKOW-UHFFFAOYSA-N | |
methyl 2-amino-5-methoxybenzoate | |
13052275 | |
96% |
2475-80-1 | |
C9H11NO3 | |
250 mg | |
Slightly Soluble (1.1g/L) (25°C) in water. | |
COC(=O)C1=C(N)C=CC(OC)=C1 | |
181.19 | |
181.19 |
Chemical Identifiers
2475-80-1 | |
181.19 | |
MOVBJUGHBJJKOW-UHFFFAOYSA-N | |
methyl 2-amino-5-methoxybenzoate |
C9H11NO3 | |
MFCD09038482 | |
13052275 | |
COC(=O)C1=C(N)C=CC(OC)=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only