missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-amino-5-methoxybenzoate, 96%

Manufacturer:  Alfa Aesar™ H5470403

 View more versions of this product

Catalog No. AAH5470403



Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly Soluble (1.1 g/L) (25°C) in water.

Notes
Store at room temperature. Keep away from oxidizing agents and acids. Moisture sensitive.
Specifications

Specifications

Methyl 2-amino-5-methoxybenzoate
1g
C9H11NO3
2-amino-5-methoxy-benzoic acid methyl ester, 2-amino-5-methoxybenzoic acid methyl ester, 2-amino-5-methoxybenzoic acid,methyl ester, acmc-1cnv1, benzoic acid, 2-amino-5-methoxy-, methyl ester, benzoic acid,2-amino-5-methoxy-, methyl ester, intermediates-zcf02665, methyl 5-methoxyanthranilate, methyl 6-amino-m-anisate, methyl2-amino-5-methoxybenzoate
MOVBJUGHBJJKOW-UHFFFAOYSA-N
methyl 2-amino-5-methoxybenzoate
13052275
96%
32°C to 33°C
2475-80-1
MFCD09038482
Slightly Soluble (1.1g/L) (25°C) in water.
COC1=CC(=C(C=C1)N)C(=O)OC
181.191
181.19

Chemical Identifiers

2475-80-1
181.191
MOVBJUGHBJJKOW-UHFFFAOYSA-N
13052275
COC1=CC(=C(C=C1)N)C(=O)OC
C9H11NO3
MFCD09038482
2-amino-5-methoxy-benzoic acid methyl ester, 2-amino-5-methoxybenzoic acid methyl ester, 2-amino-5-methoxybenzoic acid,methyl ester, acmc-1cnv1, benzoic acid, 2-amino-5-methoxy-, methyl ester, benzoic acid,2-amino-5-methoxy-, methyl ester, intermediates-zcf02665, methyl 5-methoxyanthranilate, methyl 6-amino-m-anisate, methyl2-amino-5-methoxybenzoate
methyl 2-amino-5-methoxybenzoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents