missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-amino-5-methoxybenzoate, 96%

Manufacturer:  Alfa Aesar™ H5470406

 View more versions of this product

Catalog No. AAH5470406



Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly Soluble (1.1 g/L) (25°C) in water.

Notes
Store at room temperature. Keep away from oxidizing agents and acids. Moisture sensitive.
Specifications

Specifications

Methyl 2-amino-5-methoxybenzoate
5g
C9H11NO3
2-amino-5-methoxy-benzoic acid methyl ester
COC1=CC(=C(C=C1)N)C(=O)OC
181.191
181.19
32°C to 33°C
2475-80-1
MFCD09038482
MOVBJUGHBJJKOW-UHFFFAOYSA-N
methyl 2-amino-5-methoxybenzoate
13052275
96%

Chemical Identifiers

2475-80-1
181.191
MOVBJUGHBJJKOW-UHFFFAOYSA-N
13052275
COC1=CC(=C(C=C1)N)C(=O)OC
C9H11NO3
MFCD09038482
2-amino-5-methoxy-benzoic acid methyl ester
methyl 2-amino-5-methoxybenzoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents