Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-amino-5-chloro-3-iodobenzoate, 98+%

$112.26 - $415.86

Chemical Identifiers

Chemical Name or Material Methyl 2-amino-5-chloro-3-iodobenzoate
CAS 289039-84-5
Molecular Formula C8H7ClINO2
Formula Weight 311.51
MDL Number MFCD00673017
Synonym 2-Amino-5-chloro-3-iodobenzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2018806

  SDS
alfa aesar™
L2018806
5g Each for $112.26

AAL2018814

  SDS
alfa aesar™
L2018814
25g Each for $415.86
Description & Specifications

Specifications

Formula Weight 311.51
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym 2-Amino-5-chloro-3-iodobenzoic acid methyl ester
Quantity 5g
Chemical Name or Material Methyl 2-amino-5-chloro-3-iodobenzoate
Assay ≥98%
CAS 289039-84-5
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00673017
Health Hazard 1 Warning
Melting Point 67° to 69°C
Molecular Formula C8H7ClINO2
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.