missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC450370010

 View more versions of this product

Catalog No. AC450370010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Methyl 2-amino-5, 6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate
170°C to 174°C
184174-80-9
C9H11NO2S
XUKGZPUAFGAYHC-UHFFFAOYSA-N
methyl 2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate
2818378
95%
Glass Bottle
1g
94% min. (HPLC)
2-amino-5,6-dihydro-4h-cyclopenta b thiophene-3-ca, 2-amino-5,6-dihydro-4h-cyclopenta b thiophene-3-carboxylic acid methyl ester, 4h-cyclopenta b thiophene-3-carboxylic acid, 2-amino-5,6-dihydro-, methyl, 4h-cyclopenta b thiophene-3-carboxylicacid, 2-amino-5,6-dihydro-, methyl ester, enamine_005617, methyl 2-amino-4,5,6-trihydrocyclopenta 1,2-b thiophene-3-carboxylate, methyl 2-amino-4h,5h,6h-cyclopenta b thiophene-3-carboxylate, methyl 2-amino-5,6-dihydro-4h-cyclopenta b thiophene-3-carboxylate, methyl 2-amino-5,6-dihydro-4h-cyclopenta b-thiophene-3-carboxylate, methyl 2-azanyl-5,6-dihydro-4h-cyclopenta b thiophene-3-carboxylate
COC(=O)C1=C(SC2=C1CCC2)N
197.26
197.26

Chemical Identifiers

184174-80-9
197.26
2-amino-5,6-dihydro-4h-cyclopenta b thiophene-3-ca, 2-amino-5,6-dihydro-4h-cyclopenta b thiophene-3-carboxylic acid methyl ester, 4h-cyclopenta b thiophene-3-carboxylic acid, 2-amino-5,6-dihydro-, methyl, 4h-cyclopenta b thiophene-3-carboxylicacid, 2-amino-5,6-dihydro-, methyl ester, enamine_005617, methyl 2-amino-4,5,6-trihydrocyclopenta 1,2-b thiophene-3-carboxylate, methyl 2-amino-4h,5h,6h-cyclopenta b thiophene-3-carboxylate, methyl 2-amino-5,6-dihydro-4h-cyclopenta b thiophene-3-carboxylate, methyl 2-amino-5,6-dihydro-4h-cyclopenta b-thiophene-3-carboxylate, methyl 2-azanyl-5,6-dihydro-4h-cyclopenta b thiophene-3-carboxylate
methyl 2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate
C9H11NO2S
XUKGZPUAFGAYHC-UHFFFAOYSA-N
2818378
COC(=O)C1=C(SC2=C1CCC2)N
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS Signal Word: Warning

SDS Documents