Please login to your online account to display your discounted pricing

Methyl 2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate, 95%, Acros Organics

Chemical Identifiers

Chemical Name or Material Methyl 2-amino-5, 6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate
Assay Percent Range 95%
Molecular Formula C9H11NO2S
Formula Weight 197.26
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC450370010

  SDS
thermo fisher scientific
AC450370010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC450370100

  SDS
thermo fisher scientific
AC450370100
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 197.26
Quantity 1g
Chemical Name or Material Methyl 2-amino-5, 6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate
Assay Percent Range 95%
Molecular Formula C9H11NO2S
CAS 184174-80-9