missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2-Amino-4-(trifluoromethyl)benzoate, TCI America™

$77.92 - $299.45

Chemical Identifiers

CAS 61500-87-6
Molecular Formula C9H8F3NO2
Molecular Weight (g/mol) 219.163
MDL Number MFCD08543939
InChI Key DZICUHOFOOPVFM-UHFFFAOYSA-N
Synonym 2-Amino-4-(trifluoromethyl)benzoic Acid Methyl Ester
PubChem CID 12601886
IUPAC Name methyl 2-amino-4-(trifluoromethyl)benzoate
SMILES COC(=O)C1=C(C=C(C=C1)C(F)(F)F)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M29055G View Documents TCI America
M29055G
5g Each for $299.45
 
M29051G View Documents TCI America
M29051G
1g Each for $77.92
 
Specifications

Chemical Identifiers

61500-87-6
219.163
DZICUHOFOOPVFM-UHFFFAOYSA-N
12601886
COC(=O)C1=C(C=C(C=C1)C(F)(F)F)N
C9H8F3NO2
MFCD08543939
2-Amino-4-(trifluoromethyl)benzoic Acid Methyl Ester
methyl 2-amino-4-(trifluoromethyl)benzoate

Specifications

Undesignated
5g
C9H8F3NO2
2-Amino-4-(trifluoromethyl)benzoic Acid Methyl Ester
COC(=O)C1=C(C=C(C=C1)C(F)(F)F)N
219.163
219.16
≥98.0% (GC,T)
63°C
61500-87-6
MFCD08543939
DZICUHOFOOPVFM-UHFFFAOYSA-N
methyl 2-amino-4-(trifluoromethyl)benzoate
12601886
Crystal-Powder at 20°C
Methyl 2-Amino-4-(trifluoromethyl)benzoate
Safety and Handling

Safety and Handling

TSCA : No