missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2-amino-4-fluorobenzoate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6174914

 View more versions of this product

Catalog No. AAH6174914



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methyl 2-amino-4-fluorobenzoate
C8H8FNO2
25 g
UBFRSTYHLYPSND-UHFFFAOYSA-N
methyl 2-amino-4-fluorobenzoate
14075433
98%
2475-81-2
MFCD11054082
methyl2-amino-4-fluorobenzoate, 2-amino-4-fluoro-benzoic acid methyl ester, benzoic acid, 2-amino-4-fluoro-, methyl ester, acmc-209gej, intermediates-zcf02666, methyl 4-fluoroanthranilate, 2-amino-4-fluorobenzoic acid methyl ester
COC(=O)C1=C(N)C=C(F)C=C1
169.16
169.15

Chemical Identifiers

2475-81-2
169.16
UBFRSTYHLYPSND-UHFFFAOYSA-N
14075433
COC(=O)C1=C(N)C=C(F)C=C1
C8H8FNO2
MFCD11054082
methyl2-amino-4-fluorobenzoate, 2-amino-4-fluoro-benzoic acid methyl ester, benzoic acid, 2-amino-4-fluoro-, methyl ester, acmc-209gej, intermediates-zcf02666, methyl 4-fluoroanthranilate, 2-amino-4-fluorobenzoic acid methyl ester
methyl 2-amino-4-fluorobenzoate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only