missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-amino-4,5-difluorobenzoate, 98%

$30.12 - $357.89

Chemical Identifiers

CAS 207346-42-7
Molecular Formula C8H7F2NO2
Molecular Weight (g/mol) 187.146
MDL Number MFCD09264511
InChI Key VIAQNTRKUPBQKR-UHFFFAOYSA-N
Synonym 2-amino-4,5-difluoro-benzoic acid methyl ester, 2-amino-4,5-difluorobenzoic acid methyl ester, acmc-209fdn, benzoic acid, 2-amino-4,5-difluoro-, methyl ester, ksc496g7j, methyl 2-amino-4,5-difluoro-benzoate, methyl 4,5-difluoroanthranilate, methyl2-amino-4,5-difluorobenzoate, pubchem4676
PubChem CID 18472001
IUPAC Name methyl 2-amino-4,5-difluorobenzoate
SMILES COC(=O)C1=CC(=C(C=C1N)F)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6438506 View Documents Alfa Aesar™
H6438506
5g Each for $30.12
 
AAH6438514 View Documents Alfa Aesar™
H6438514
25g Each for $98.05
 
AAH6438522 View Documents Alfa Aesar™
H6438522
100g Each for $357.89
 
Specifications

Chemical Identifiers

207346-42-7
187.146
VIAQNTRKUPBQKR-UHFFFAOYSA-N
18472001
COC(=O)C1=CC(=C(C=C1N)F)F
C8H7F2NO2
MFCD09264511
2-amino-4,5-difluoro-benzoic acid methyl ester, 2-amino-4,5-difluorobenzoic acid methyl ester, acmc-209fdn, benzoic acid, 2-amino-4,5-difluoro-, methyl ester, ksc496g7j, methyl 2-amino-4,5-difluoro-benzoate, methyl 4,5-difluoroanthranilate, methyl2-amino-4,5-difluorobenzoate, pubchem4676
methyl 2-amino-4,5-difluorobenzoate

Specifications

White
207346-42-7
MFCD09264511
2-amino-4,5-difluoro-benzoic acid methyl ester, 2-amino-4,5-difluorobenzoic acid methyl ester, acmc-209fdn, benzoic acid, 2-amino-4,5-difluoro-, methyl ester, ksc496g7j, methyl 2-amino-4,5-difluoro-benzoate, methyl 4,5-difluoroanthranilate, methyl2-amino-4,5-difluorobenzoate, pubchem4676
COC(=O)C1=CC(=C(C=C1N)F)F
187.146
187.15
98%
5g
C8H7F2NO2
2834936
VIAQNTRKUPBQKR-UHFFFAOYSA-N
methyl 2-amino-4,5-difluorobenzoate
18472001
Solid
Methyl 2-amino-4,5-difluorobenzoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures