missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-amino-3-methylbenzoate, 98%

Manufacturer:  Alfa Aesar™ H6144822

 View more versions of this product

Catalog No. AAH6144822



Specifications

Specifications

Methyl 2-amino-3-methylbenzoate
Pink
100g
C9H11NO2
2-amino-3-methyl-benzoic acid methyl ester, 2-amino-3-methylbenzoic acid methyl ester, acmc-209ftn, benzoic acid, 2-amino-3-methyl-, methyl ester, ksc495o6t, methyl 2-amino-3-methylbenzenecarboxylate, methyl 2-amino-m-toluate, methyl 3-methylanthranilate, methyl2-amino-3-methylbenzoate, pubchem10929
CC1=CC=CC(=C1N)C(=O)OC
165.192
165.19
98%
153°C (23 mm)
25°C to 29°C
22223-49-0
MFCD06200918
VSFYTPXXMLJNAU-UHFFFAOYSA-N
methyl 2-amino-3-methylbenzoate
2763406
Solid

Chemical Identifiers

22223-49-0
165.192
VSFYTPXXMLJNAU-UHFFFAOYSA-N
2763406
CC1=CC=CC(=C1N)C(=O)OC
C9H11NO2
MFCD06200918
2-amino-3-methyl-benzoic acid methyl ester, 2-amino-3-methylbenzoic acid methyl ester, acmc-209ftn, benzoic acid, 2-amino-3-methyl-, methyl ester, ksc495o6t, methyl 2-amino-3-methylbenzenecarboxylate, methyl 2-amino-m-toluate, methyl 3-methylanthranilate, methyl2-amino-3-methylbenzoate, pubchem10929
methyl 2-amino-3-methylbenzoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 244-847-6

TSCA : No

Recommended Storage : Ambient temperatures

Documents