missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2-Amino-3-methoxybenzoate, TCI America™

$163.47 - $484.31

Chemical Identifiers

CAS 5121-34-6
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.191
MDL Number MFCD11113109
InChI Key YJEZEMGLLFLMDF-UHFFFAOYSA-N
Synonym 2-Amino-3-methoxybenzoic Acid Methyl Ester, Methyl 2-Amino-m-anisate, 2-Amino-m-anisic Acid Methyl Ester
PubChem CID 602320
IUPAC Name methyl 2-amino-3-methoxybenzoate
SMILES COC1=CC=CC(=C1N)C(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M28225G View Documents TCI America
M28225G
5g Each for $484.31
 
M28221G View Documents TCI America
M28221G
1g Each for $163.47
 
Specifications

Chemical Identifiers

5121-34-6
181.191
YJEZEMGLLFLMDF-UHFFFAOYSA-N
602320
COC1=CC=CC(=C1N)C(=O)OC
C9H11NO3
MFCD11113109
2-Amino-3-methoxybenzoic Acid Methyl Ester, Methyl 2-Amino-m-anisate, 2-Amino-m-anisic Acid Methyl Ester
methyl 2-amino-3-methoxybenzoate

Specifications

117°C
48°C
5121-34-6
MFCD11113109
YJEZEMGLLFLMDF-UHFFFAOYSA-N
methyl 2-amino-3-methoxybenzoate
602320
Crystal-Powder at 20°C
Methyl 2-Amino-3-methoxybenzoate
Undesignated
5g
C9H11NO3
2-Amino-3-methoxybenzoic Acid Methyl Ester, Methyl 2-Amino-m-anisate, 2-Amino-m-anisic Acid Methyl Ester
COC1=CC=CC(=C1N)C(=O)OC
181.191
181.19
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : No