Please login to your online account to display your discounted pricing

Methyl 2-amino-3-carboxybenzoate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Methyl 2-amino-3-carboxybenzoate
CAS 253120-47-7
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C9H9NO4
Formula Weight 195.17
MDL Number MFCD06200897
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO07276CB

  SDS
fisher scientific company
MO07276CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 195.17
Quantity 250mg
Assay 97%
CAS Min % 97.0
Chemical Name or Material Methyl 2-amino-3-carboxybenzoate
Molecular Formula C9H9NO4
MDL Number MFCD06200897
CAS 253120-47-7
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber Glass Bottle
Assay Percent Range 97%