missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-amino-3-bromo-5-methoxybenzoate, 96%

Manufacturer:  Alfa Aesar™ H5484403

 View more versions of this product

Catalog No. AAH5484403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

Methyl 2-amino-3-bromo-5-methoxybenzoate
1378874-22-6
MFCD20527215
WPRSGBOUSTUNEC-UHFFFAOYSA-N
methyl 2-amino-3-bromo-5-methoxybenzoate
73995991
96%
1g
C9H10BrNO3
2-amino-3-bromo-5-methoxybenzoic acid methyl ester
COC1=CC(=C(C(=C1)C(=O)OC)N)Br
260.087
260.09

Chemical Identifiers

1378874-22-6
260.087
WPRSGBOUSTUNEC-UHFFFAOYSA-N
73995991
COC1=CC(=C(C(=C1)C(=O)OC)N)Br
C9H10BrNO3
MFCD20527215
2-amino-3-bromo-5-methoxybenzoic acid methyl ester
methyl 2-amino-3-bromo-5-methoxybenzoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents