Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-amino-3,5-dibromobenzoate, 98+%

$24.49 - $109.15

Chemical Identifiers

Chemical Name or Material Methyl 2-amino-3,5-dibromobenzoate
CAS 606-00-8
Molecular Formula C8H7Br2NO2
Formula Weight 308.97
MDL Number MFCD00010873
Beilstein 3257505
Synonym 2-Amino-3,5-dibromobenzoic acid methyl ester; Methyl 3,5-dibromoanthranilate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2251806

 
alfa aesar™
B2251806
5g Each for $24.49

AAB2251822

 
alfa aesar™
B2251822
100g Each for $109.15
Description & Specifications

Specifications

Formula Weight 308.97
Chemical Name or Material Methyl 2-amino-3,5-dibromobenzoate
TSCA TSCA
Assay ≥98%
Molecular Formula C8H7Br2NO2
MDL Number MFCD00010873
CAS 606-00-8
Beilstein 3257505
Synonym 2-Amino-3,5-dibromobenzoic acid methyl ester; Methyl 3,5-dibromoanthranilate
Health Hazard 1 Warning
Melting Point 87° to 89°C
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.