Please login to your online account to display your discounted pricing

Methyl 2-(acetylamino)-3-nitrobenzoate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material Methyl 2-(acetylamino)-3-nitrobenzoate
CAS 95067-27-9
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H10N2O5
Formula Weight 238.2
MDL Number MFCD00157613
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RH00481DA

  SDS
fisher scientific company
RH00481DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 238.2
Quantity 1g
Assay 97%
CAS Min % 97.0
Chemical Name or Material Methyl 2-(acetylamino)-3-nitrobenzoate
Molecular Formula C10H10N2O5
MDL Number MFCD00157613
Packaging Amber Glass Bottle
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 95067-27-9
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.