Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-acetamido-5-chlorobenzoate, 97%

$314.01 - $314.01

Chemical Identifiers

Chemical Name or Material Methyl 2-acetamido-5-chlorobenzoate
CAS 20676-54-4
Molecular Formula C10H10ClNO3
Formula Weight 227.65
MDL Number MFCD00144759
Beilstein 2808490
Synonym 2-Acetamido-5-chlorobenzoic acid methyl ester; N-Acetyl-5-chloroanthranilic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1682630

 
alfa aesar™
A1682630
250g Each for $314.01
Description & Specifications

Specifications

Formula Weight 227.65
Chemical Name or Material Methyl 2-acetamido-5-chlorobenzoate
Assay 97%
Molecular Formula C10H10ClNO3
MDL Number MFCD00144759
CAS 20676-54-4
Beilstein 2808490
Synonym 2-Acetamido-5-chlorobenzoic acid methyl ester; N-Acetyl-5-chloroanthranilic acid methyl ester
Melting Point 128° to 131°C
Quantity 250g