Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-acetamido-5-bromobenzoate, 97%

$75.57 - $224.73

Chemical Identifiers

Chemical Name or Material Methyl 2-acetamido-5-bromobenzoate
CAS 138825-96-4
Molecular Formula C10H10BrNO3
Formula Weight 272.1
MDL Number MFCD00144758
Beilstein 7810898
Synonym 2-Acetamido-5-bromobenzoic acid methyl ester; N-Acetyl-5-bromoanthranilic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1708714

  SDS
alfa aesar™
A1708714
25g Each for $75.57

AAA1708722

  SDS
alfa aesar™
A1708722
100g Each for $224.73
Description & Specifications

Specifications

Formula Weight 272.1
Chemical Name or Material Methyl 2-acetamido-5-bromobenzoate
Assay 97%
Molecular Formula C10H10BrNO3
MDL Number MFCD00144758
CAS 138825-96-4
Beilstein 7810898
Synonym 2-Acetamido-5-bromobenzoic acid methyl ester; N-Acetyl-5-bromoanthranilic acid methyl ester
Melting Point 132° to 135°C
Quantity 25g