missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 2,6-dihydroxybenzoate, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC442450010
Specifications
Methyl 2, 6-dihydroxybenzoate | |
67°C to 69°C | |
2150-45-0 | |
C8H8O4 | |
WCQZCKUNZVMBDC-UHFFFAOYSA-N | |
methyl 2,6-dihydroxybenzoate | |
519869 | |
97% |
Glass Bottle | |
1g | |
96% min. (GC) | |
.gamma.-resorcylic acid, methyl ester, 2,6-dihydroxybenzoic acid methyl ester, 2-methoxycarbonyl benzene-1,3-diol, 2-methoxycarbonyl resorcinol, acmc-209fl9, benzoic acid, 2,6-dihydroxy-, methyl ester, gamma-resorcylic acid methyl ester, methyl 2,6-dihydroxy benzoate, methyl 2,6-dihydroxybenzoate, methyl-2,6-dihydroxybenzoate | |
COC(=O)C1=C(C=CC=C1O)O | |
168.15 | |
168.15 |
Chemical Identifiers
2150-45-0 | |
168.15 | |
.gamma.-resorcylic acid, methyl ester, 2,6-dihydroxybenzoic acid methyl ester, 2-methoxycarbonyl benzene-1,3-diol, 2-methoxycarbonyl resorcinol, acmc-209fl9, benzoic acid, 2,6-dihydroxy-, methyl ester, gamma-resorcylic acid methyl ester, methyl 2,6-dihydroxy benzoate, methyl 2,6-dihydroxybenzoate, methyl-2,6-dihydroxybenzoate | |
methyl 2,6-dihydroxybenzoate |
C8H8O4 | |
WCQZCKUNZVMBDC-UHFFFAOYSA-N | |
519869 | |
COC(=O)C1=C(C=CC=C1O)O |
Safety and Handling
GHS Signal Word: Warning