Please login to your online account to display your discounted pricing

Methyl 2,6-dihydroxybenzoate, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material Methyl 2, 6-dihydroxybenzoate
Assay Percent Range 97%
Molecular Formula C8H8O4
Formula Weight 168.15
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC442450010

 
thermo fisher scientific
AC442450010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC442450050

 
thermo fisher scientific
AC442450050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 168.15
Quantity 1g
Molecular Formula C8H8O4
Chemical Name or Material Methyl 2, 6-dihydroxybenzoate
CAS 2150-45-0
Assay Percent Range 97%