missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2,6-dihydroxybenzoate, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC442450050

 View more versions of this product

Catalog No. AC442450050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Methyl 2, 6-dihydroxybenzoate
5g
96% min. (GC)
.gamma.-resorcylic acid, methyl ester, 2,6-dihydroxybenzoic acid methyl ester, 2-methoxycarbonyl benzene-1,3-diol, 2-methoxycarbonyl resorcinol, acmc-209fl9, benzoic acid, 2,6-dihydroxy-, methyl ester, gamma-resorcylic acid methyl ester, methyl 2,6-dihydroxy benzoate, methyl 2,6-dihydroxybenzoate, methyl-2,6-dihydroxybenzoate
COC(=O)C1=C(C=CC=C1O)O
168.15
168.15
67°C to 69°C
2150-45-0
C8H8O4
WCQZCKUNZVMBDC-UHFFFAOYSA-N
methyl 2,6-dihydroxybenzoate
519869
97%

Chemical Identifiers

2150-45-0
168.15
.gamma.-resorcylic acid, methyl ester, 2,6-dihydroxybenzoic acid methyl ester, 2-methoxycarbonyl benzene-1,3-diol, 2-methoxycarbonyl resorcinol, acmc-209fl9, benzoic acid, 2,6-dihydroxy-, methyl ester, gamma-resorcylic acid methyl ester, methyl 2,6-dihydroxy benzoate, methyl 2,6-dihydroxybenzoate, methyl-2,6-dihydroxybenzoate
methyl 2,6-dihydroxybenzoate
C8H8O4
WCQZCKUNZVMBDC-UHFFFAOYSA-N
519869
COC(=O)C1=C(C=CC=C1O)O
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents