missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2,6-difluoro-3-nitrobenzoate, 98%

$30.86 - $495.01

Chemical Identifiers

CAS 84832-01-9
Molecular Formula C8H5F2NO4
Molecular Weight (g/mol) 217.128
MDL Number MFCD07368850
InChI Key CIHHBTMZOLRCRL-UHFFFAOYSA-N
Synonym 2,6-difluoro-3-nitro-benzoic acid methyl ester
PubChem CID 43449035
IUPAC Name methyl 2,6-difluoro-3-nitrobenzoate
SMILES COC(=O)C1=C(C=CC(=C1F)[N+](=O)[O-])F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6132703 View Documents Alfa Aesar™
H6132703
1g Each for $30.86
 
AAH6132706 View Documents Alfa Aesar™
H6132706
5g Each for $112.82
 
AAH6132714 View Documents Alfa Aesar™
H6132714
25g Each for $495.01
 
Specifications

Chemical Identifiers

84832-01-9
217.128
CIHHBTMZOLRCRL-UHFFFAOYSA-N
43449035
COC(=O)C1=C(C=CC(=C1F)[N+](=O)[O-])F
C8H5F2NO4
MFCD07368850
2,6-difluoro-3-nitro-benzoic acid methyl ester
methyl 2,6-difluoro-3-nitrobenzoate

Specifications

1g
C8H5F2NO4
2,6-difluoro-3-nitro-benzoic acid methyl ester
COC(=O)C1=C(C=CC(=C1F)[N+](=O)[O-])F
217.128
217.13
98%
84832-01-9
MFCD07368850
CIHHBTMZOLRCRL-UHFFFAOYSA-N
methyl 2,6-difluoro-3-nitrobenzoate
43449035
Solid
Methyl 2,6-difluoro-3-nitrobenzoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures