Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2,6-dichloronicotinate, 97%

$93.42 - $254.65

Chemical Identifiers

Chemical Name or Material Methyl 2,6-dichloronicotinate
CAS 65515-28-8
Molecular Formula C7H5Cl2NO2
Formula Weight 206.03
MDL Number MFCD07369794
Synonym 2,6-Dichloronicotinic acid methyl ester; Methyl 2,6-dichloropyridine-3-carboxylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5109203

  SDS
alfa aesar™
H5109203
1g Each for $254.65

AAH51092MD

  SDS
alfa aesar™
H51092MD
250mg Each for $93.42
Description & Specifications

Specifications

Formula Weight 206.03
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Chemical Name or Material Methyl 2,6-dichloronicotinate
CAS 65515-28-8
Synonym 2,6-Dichloronicotinic acid methyl ester; Methyl 2,6-dichloropyridine-3-carboxylate
MDL Number MFCD07369794
Molecular Formula C7H5Cl2NO2
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.