Please login to your online account to display your discounted pricing

Methyl 2,6-dichloro-5-fluoronicotinate, 90%, Maybridge

Chemical Identifiers

Chemical Name or Material Methyl 2, 6-dichloro-5-fluoronicotinate
CAS 189281-66-1
CAS Min % 90.0
Assay 90%
Assay Percent Range 90%
Molecular Formula C7H4Cl2FNO2
Formula Weight 224.02
MDL Number MFCD02180634
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RH02135EA

 
fisher scientific company
RH02135EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 224.02
Quantity 10g
Assay 90%
CAS Min % 90.0
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber Glass Bottle
Assay Percent Range 90%
MDL Number MFCD02180634
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Molecular Formula C7H4Cl2FNO2
CAS 189281-66-1
Chemical Name or Material Methyl 2, 6-dichloro-5-fluoronicotinate