Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2,5-dimethylpyrrole-3-carboxylate, 97%

$256.15 - $256.15

Chemical Identifiers

Chemical Name or Material Methyl 2,5-dimethylpyrrole-3-carboxylate
CAS 69687-80-5
Molecular Formula C8H11NO2
Formula Weight 153.18
MDL Number MFCD00203859
Synonym 2,5-Dimethylpyrrole-3-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1998006

  SDS
alfa aesar™
A1998006
5g Each for $256.15
Description & Specifications

Specifications

Formula Weight 153.18
Chemical Name or Material Methyl 2,5-dimethylpyrrole-3-carboxylate
MDL Number MFCD00203859
Assay 97%
Molecular Formula C8H11NO2
CAS 69687-80-5
Synonym 2,5-Dimethylpyrrole-3-carboxylic acid methyl ester
Melting Point 118° to 122°C
Quantity 5g